Gene Transcripts

Gene Symbol
ANKS1A
Entrez Gene ID
23294
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1A
Gene Aliases
ANKS1
Chromosome Location
Chr. 6: 34889261 - 35097994 on Build GRCh38
UniGene ID
Hs.656492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015245.2 NP_056060.2 10 1510
XM_005248964.3 XP_005249021.1 11 1561
XM_006715036.2 XP_006715099.1 11 1561
XM_011514431.2 XP_011512733.1 11 1561
XM_011514432.2 XP_011512734.1 11 1561
XM_011514433.1 XP_011512735.1 11 1561
XM_011514434.2 XP_011512736.1 10 1498
XM_011514435.2 XP_011512737.1 10 1498
XM_011514436.2 XP_011512738.1 11 5130
GenBank mRNA BC022396.1 - 10 1510
BC031934.1 - 11 1582 4819948
BC132832.1 - 10 1447 40146854
D86982.1 - 10 1507

View Full Product Details