Gene Transcripts

Gene Symbol
Lsm6
Entrez Gene ID
498934
Gene Name
LSM6 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
RGD1561937
Chromosome Location
Chr. 19: 32770643 - 32785215 on Build Rnor_6.0
UniGene ID
Rn.139376
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126085.1 NP_001119557.1 4 323
XM_006255408.1 XP_006255470.1 3 238

Gene Symbol
LOC100360750
Entrez Gene ID
100360750
Gene Name
Sm protein F-like
Gene Aliases
Chromosome Location
Chr. 13: 47380384 - 47382933 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002728002.4 XP_002728048.1 1 296

View Full Product Details