Gene Transcripts
- Gene Symbol
- FKBP15
- Entrez Gene ID
- 23307
- Gene Name
- FK506 binding protein 15
- Gene Aliases
- FKBP133, KIAA0674, PPP1R76
- Chromosome Location
- Chr. 9: 113165036 - 113221361 on Build GRCh38
- UniGene ID
- Hs.522351
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015258.1 |
NP_056073.1
|
5, 6 |
525 |
|
XM_006717018.2 |
XP_006717081.1
|
5, 6 |
442 |
|
XM_006717019.1 |
XP_006717082.1
|
4, 5 |
373 |
|
XM_017014552.1 |
XP_016870041.1
|
4, 5 |
373 |
|
GenBank mRNA |
AB014574.1 |
- |
5, 6 |
452 |
|
AK303074.1 |
- |
4, 5 |
373 |
|
AK307498.1 |
- |
4, 5 |
354 |
|
AK307507.1 |
- |
9, 10 |
959 |
|
BC009609.1 |
- |
5, 6 |
482 |
|
DC321217.1 |
- |
5, 6 |
526 |
|
View Full Product Details