Gene Transcripts

Gene Symbol
Wbscr27
Entrez Gene ID
688407
Gene Name
Williams Beuren syndrome chromosome region 27
Gene Aliases
Chromosome Location
Chr. 12: 24766986 - 24776291 on Build Rnor_6.0
UniGene ID
Rn.163207
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109499.2 NP_001102969.1 5 630
XM_006249190.2 XP_006249252.1 6 716
XM_006249191.2 XP_006249253.1 6 716

View Full Product Details