Gene Transcripts

Gene Symbol
Chst14
Entrez Gene ID
691394
Gene Name
carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 14
Gene Aliases
Chromosome Location
Chr. 3: 110734105 - 110736162 on Build Rnor_6.0
UniGene ID
Rn.106173
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109639.1 NP_001103109.1 1 927
GenBank mRNA BC166542.1 - 1 872

View Full Product Details