Gene Transcripts

Gene Symbol
Gng8
Entrez Gene ID
245986
Gene Name
guanine nucleotide binding protein (G protein), gamma 8
Gene Aliases
Chromosome Location
Chr. 1: 78818377 - 78822224 on Build Rnor_6.0
UniGene ID
Rn.11233
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139185.1 NP_631924.1 4 472
XM_006228384.2 XP_006228446.1 3 826
XM_006228385.2 XP_006228447.1 3 270
GenBank mRNA L35921.1 - 4 472

View Full Product Details