Gene Transcripts

Gene Symbol
Sprn
Entrez Gene ID
541462
Gene Name
shadow of prion protein homolog (zebrafish)
Gene Aliases
Chromosome Location
Chr. 1: 212619817 - 212623681 on Build Rnor_6.0
UniGene ID
Rn.23342
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031845.1 NP_001027015.1 1 383
XM_006230573.2 XP_006230635.1 2 660

View Full Product Details