Gene Transcripts

Gene Symbol
Aurkc
Entrez Gene ID
292554
Gene Name
aurora kinase C
Gene Aliases
Chromosome Location
Chr. 1: 69958452 - 69964105 on Build Rnor_6.0
UniGene ID
Rn.41092
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106221.1 NP_001099691.1 3 565
XM_008758900.1 XP_008757122.1 6 929
XM_008758901.1 XP_008757123.1 7 1032

View Full Product Details