Gene Transcripts

Gene Symbol
Syndig1l
Entrez Gene ID
685882
Gene Name
synapse differentiation inducing 1-like
Gene Aliases
Tmem90a
Chromosome Location
Chr. 6: 108392837 - 108415096 on Build Rnor_6.0
UniGene ID
Rn.171669
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109485.1 NP_001102955.1 3 562
XM_006240353.2 XP_006240415.1 2 464

View Full Product Details