Gene Transcripts

Gene Symbol
Polr2h
Entrez Gene ID
498109
Gene Name
polymerase (RNA) II (DNA directed) polypeptide H
Gene Aliases
RGD1561203, RPB17
Chromosome Location
Chr. 11: 83877760 - 83882208 on Build Rnor_6.0
UniGene ID
Rn.9160
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134789.1 NP_001128261.1 2 237
GenBank mRNA CB758009.1 - 2 251
CF113682.1 - 2 210
FQ222889.1 - 2 235

Gene Symbol
LOC100912534
Entrez Gene ID
100912534
Gene Name
DNA-directed RNA polymerases I, II, and III subunit RPABC3-like
Gene Aliases
Chromosome Location
Chr. 11: 82856537 - 82860991 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751083.2 XP_003751131.1 2 251

View Full Product Details