Gene Transcripts

Gene Symbol
Sbk2
Entrez Gene ID
691411
Gene Name
SH3 domain binding kinase family, member 2
Gene Aliases
RGD1563279
Chromosome Location
Chr. 1: 72425728 - 72433494 on Build Rnor_6.0
UniGene ID
Rn.201731
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127539.2 NP_001121011.1 4 2307
GenBank mRNA BC161893.1 - 4 2307

View Full Product Details