Gene Transcripts

Gene Symbol
Leap2
Entrez Gene ID
497901
Gene Name
liver-expressed antimicrobial peptide 2
Gene Aliases
Ac1177
Chromosome Location
Chr. 10: 38771125 - 38774515 on Build Rnor_6.0
UniGene ID
Rn.198854
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126081.1 NP_001119553.1 3 137
GenBank mRNA AY310147.1 - 3 137

View Full Product Details