Gene Transcripts

Gene Symbol
Aldh4a1
Entrez Gene ID
641316
Gene Name
aldehyde dehydrogenase 4 family, member A1
Gene Aliases
Chromosome Location
Chr. 5: 158090238 - 158115725 on Build Rnor_6.0
UniGene ID
Rn.203318
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134698.1 NP_001128170.1 15 3107
GenBank mRNA BC168153.1 - 15 3107

View Full Product Details