Gene Transcripts

Gene Symbol
Ttll13
Entrez Gene ID
308762
Gene Name
tubulin tyrosine ligase-like family, member 13
Gene Aliases
RGD1310399
Chromosome Location
Chr. 1: 142034423 - 142050071 on Build Rnor_6.0
UniGene ID
Rn.186496
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134962.1 NP_001128434.1 1 247
XM_008759532.1 XP_008757754.1 1 247
GenBank mRNA BC169015.1 - 1 247

View Full Product Details