Gene Transcripts

Gene Symbol
Rup2
Entrez Gene ID
619560
Gene Name
urinary protein 2
Gene Aliases
UP-1
Chromosome Location
Chr. 8: 41657662 - 41661223 on Build Rnor_6.0
UniGene ID
Rn.144564
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034950.2 NP_001030122.2 2, 3 233
GenBank mRNA AF198441.1 - 2, 3 205
AY327506.1 - 2, 3 233

View Full Product Details