Gene Transcripts
- Gene Symbol
- TRAM1
- Entrez Gene ID
- 23471
- Gene Name
- translocation associated membrane protein 1
- Gene Aliases
- PNAS8, TRAM, TRAMP
- Chromosome Location
- Chr. 8: 70573218 - 70608459 on Build GRCh38
- UniGene ID
- Hs.491988
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317804.1 |
NP_001304733.1
|
8, 9 |
824 |
|
NM_001317805.1 |
NP_001304734.1
|
7, 8 |
1339 |
|
NM_014294.5 |
NP_055109.1
|
7, 8 |
907 |
|
XM_017013266.1 |
XP_016868755.1
|
6, 7 |
779 |
|
XM_017013267.1 |
XP_016868756.1
|
6, 7 |
1123 |
|
GenBank mRNA |
AK289397.1 |
- |
7, 8 |
767 |
|
AK292777.1 |
- |
7, 8 |
691 |
|
AK303411.1 |
- |
8, 9 |
752 |
|
BC000687.2 |
- |
7, 8 |
729 |
3347823 |
BC020283.1 |
- |
7, 8 |
722 |
|
BC037738.1 |
- |
7, 8 |
819 |
5261490 |
BT007359.1 |
- |
7, 8 |
647 |
|
CR541654.1 |
- |
7, 8 |
647 |
|
CR541739.1 |
- |
7, 8 |
647 |
|
X63679.1 |
- |
7, 8 |
768 |
|
View Full Product Details