Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006135.2 | NP_006126.1 | 4 | 591 | |
XM_011542225.2 | XP_011540527.1 | 4 | 335 | ||
XM_017002424.1 | XP_016857913.1 | 4 | 335 | ||
GenBank mRNA | AK223226.1 | - | 4 | 212 | |
AK289654.1 | - | 4 | 234 | ||
BC000144.1 | - | 4 | 216 | 3047937 | |
BT019364.1 | - | 4 | 199 | ||
BX648738.1 | - | 4 | 882 | ||
CR407657.1 | - | 4 | 199 | ||
CR541819.1 | - | 4 | 199 | ||
U56637.1 | - | 4 | 199 |