Gene Transcripts
- Gene Symbol
- CCNDBP1
- Entrez Gene ID
- 23582
- Gene Name
- cyclin D1 binding protein 1
- Gene Aliases
- DIP1, GCIP, HHM
- Chromosome Location
- Chr. 15: 43185118 - 43197177 on Build GRCh38
- UniGene ID
- Hs.36794
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012142.4 |
NP_036274.3
|
9, 10 |
1162 |
|
NR_027513.2 |
|
7, 8 |
985 |
|
NR_027514.2 |
|
8, 9 |
1065 |
|
NR_045998.1 |
|
8, 9 |
1082 |
|
NR_045999.1 |
|
6, 7 |
925 |
|
XM_006720448.1 |
XP_006720511.1
|
7, 8 |
1084 |
|
GenBank mRNA |
AF082569.1 |
- |
9, 10 |
1021 |
|
AF087852.1 |
- |
9, 10 |
1007 |
|
AF113535.1 |
- |
7, 8 |
971 |
|
AF132034.1 |
- |
9, 10 |
977 |
|
AF246144.1 |
- |
9, 10 |
979 |
|
AK225143.1 |
- |
9, 10 |
977 |
|
AK225645.1 |
- |
9, 10 |
991 |
|
AK290665.1 |
- |
9, 10 |
991 |
|
AK290707.1 |
- |
8, 9 |
944 |
|
AK295017.1 |
- |
8, 9 |
894 |
|
BC009689.1 |
- |
9, 10 |
1005 |
3896421 |
BQ439439.1 |
- |
6, 7 |
739 |
|
CR450331.1 |
- |
9, 10 |
931 |
|
View Full Product Details