Gene Transcripts

Gene Symbol
SSBP2
Entrez Gene ID
23635
Gene Name
single stranded DNA binding protein 2
Gene Aliases
HSPC116, SOSS-B2
Chromosome Location
Chr. 5: 81417360 - 81751253 on Build GRCh38
UniGene ID
Hs.102735
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256732.1 NP_001243661.1 7 699
NM_001256733.1 NP_001243662.1 6 639
NM_001256734.1 NP_001243663.1 6 609
NM_001256735.1 NP_001243664.1 6 609
NM_001256736.1 NP_001243665.1 6 609
NM_012446.3 NP_036578.2 7 699
XM_011543297.2 XP_011541599.1 7 614
XM_017009304.1 XP_016864793.1 7 517
XM_017009305.1 XP_016864794.1 8 630
XM_017009306.1 XP_016864795.1 7 614
XM_017009307.1 XP_016864796.1 6 427
XM_017009308.1 XP_016864797.1 8 778
XM_017009309.1 XP_016864798.1 7 842
XM_017009310.1 XP_016864799.1 7 990
GenBank mRNA AF077048.1 - 7 536
AF161465.1 - 7 529
AK074537.1 - 7 494
AK293544.1 - 6 434
AK294864.1 - 6 404
AK297494.1 - 7 407
AK299858.1 - 6 453
AK312337.1 - 7 543
AL080076.1 - 7 547
BC017020.1 - 7 547 3913337
BI517313.1 - 7 699
CR936787.1 - 6 454

View Full Product Details