Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014331.3 | NP_055146.1 | 4 | 913 | |
XM_011531802.2 | XP_011530104.1 | 4 | 982 | ||
GenBank mRNA | AB026891.1 | - | 4 | 868 | |
AB040875.1 | - | 4 | 768 | ||
AF200708.1 | - | 4 | 878 | ||
AF252872.1 | - | 4 | 864 | ||
AJ277882.1 | - | 4 | 633 | ||
AK290359.1 | - | 4 | 913 | ||
AK314855.1 | - | 4 | 807 | ||
BC012087.1 | - | 4 | 793 | 4562994 |