Gene Transcripts
- Gene Symbol
- NPAP1
- Entrez Gene ID
- 23742
- Gene Name
- nuclear pore associated protein 1
- Gene Aliases
- C15orf2
- Chromosome Location
- Chr. 15: 24675394 - 24683446 on Build GRCh38
- UniGene ID
- Hs.649663
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018958.2 |
NP_061831.2
|
1 |
3321 |
|
View Full Product Details