Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012134.2 | NP_036266.2 | 2 | 1113 | |
GenBank mRNA | AK126474.1 | - | 2 | 652 | |
AK295824.1 | - | 2 | 1019 | ||
AK304700.1 | - | 2 | 730 | ||
AK304852.1 | - | 3 | 879 | ||
AK311740.1 | - | 3 | 977 | ||
BC080187.1 | - | 2 | 1062 | 6177902 | |
X54162.1 | - | 2 | 993 |