Gene Transcripts

Gene Symbol
LMOD1
Entrez Gene ID
25802
Gene Name
leiomodin 1
Gene Aliases
1D, 64kD, D1, SM-LMOD, SMLMOD
Chromosome Location
Chr. 1: 201896456 - 201946588 on Build GRCh38
UniGene ID
Hs.519075
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012134.2 NP_036266.2 2 747
GenBank mRNA AK295824.1 - 2 653
AK304700.1 - 2 364
AK304852.1 - 3 513
AK311740.1 - 3 611
BC001755.1 - 2 628
BC080187.1 - 2 696 6177902
X54162.1 - 2 627

View Full Product Details