Gene Transcripts

Gene Symbol
KLK5
Entrez Gene ID
25818
Gene Name
kallikrein related peptidase 5
Gene Aliases
KLK-L2, KLKL2, SCTE
Chromosome Location
Chr. 19: 50943299 - 50953088 on Build GRCh38
UniGene ID
Hs.50915
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077491.1 NP_001070959.1 5 697
NM_001077492.1 NP_001070960.1 4 629
NM_012427.4 NP_036559.1 4 832
XM_011526702.1 XP_011525004.1 4 571
XM_011526703.2 XP_011525005.1 5 651
GenBank mRNA AF168768.2 - 4 648
AF435980.1 - 4 803
AF435981.1 - 4 600
AK223421.1 - 4 780
AY279380.1 - 4 606
AY279381.1 - 5 674
AY359010.1 - 4 832
AY461805.1 - 3 490
BC008036.2 - 4 763 2989806
BT006867.1 - 3 479

View Full Product Details