Gene Transcripts

Gene Symbol
KBTBD2
Entrez Gene ID
25948
Gene Name
kelch repeat and BTB domain containing 2
Gene Aliases
BKLHD1
Chromosome Location
Chr. 7: 32868166 - 32892144 on Build GRCh38
UniGene ID
Hs.372541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015483.2 NP_056298.2 4 1582
XM_005249696.2 XP_005249753.1 4 1438
XM_005249697.2 XP_005249754.1 4 1171
XM_011515256.2 XP_011513558.1 4 1317
XM_011515257.2 XP_011513559.1 4 1051
GenBank mRNA AB040922.1 - 1 2206
AF151831.1 - 3 991
AF161402.1 - 1 342
AK024557.1 - 1 547
AK292802.1 - 4 1304
AL117562.1 - 1 100
BC037887.2 - 4 1486 5274706
BC047107.1 - 4 1319 5275217

View Full Product Details