Gene Transcripts
- Gene Symbol
- UPF2
- Entrez Gene ID
- 26019
- Gene Name
- UPF2 regulator of nonsense transcripts homolog (yeast)
- Gene Aliases
- HUPF2, RENT2, smg-3
- Chromosome Location
- Chr. 10: 11920022 - 12043170 on Build GRCh38
- UniGene ID
- Hs.370689
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015542.3 |
NP_056357.1
|
18, 19 |
3491 |
|
NM_080599.2 |
NP_542166.1
|
18, 19 |
3661 |
|
XM_017016034.1 |
XP_016871523.1
|
17, 18 |
3147 |
|
GenBank mRNA |
AB037829.1 |
- |
17, 18 |
3860 |
|
AF301013.1 |
- |
18, 19 |
3484 |
|
AF318574.1 |
- |
18, 19 |
3461 |
|
AK000764.1 |
- |
3, 4 |
322 |
|
AK074375.1 |
- |
3, 4 |
254 |
|
AK295209.1 |
- |
5, 6 |
560 |
|
AL080198.1 |
- |
12, 13 |
1661 |
|
AY013249.1 |
- |
17, 18 |
3410 |
|
BC114964.1 |
- |
18, 19 |
3455 |
40082739 |
BC115737.1 |
- |
18, 19 |
3455 |
40082728 |
View Full Product Details