Gene Transcripts
- Gene Symbol
- CNTNAP2
- Entrez Gene ID
- 26047
- Gene Name
- contactin associated protein-like 2
- Gene Aliases
- AUTS15, CASPR2, CDFE, NRXN4, PTHSL1
- Chromosome Location
- Chr. 7: 146116035 - 148420998 on Build GRCh38
- UniGene ID
- Hs.655684
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014141.5 |
NP_054860.1
|
20, 21 |
3892 |
|
GenBank mRNA |
AB020675.1 |
- |
20, 21 |
3401 |
|
AB073883.1 |
- |
8, 9 |
1312 |
|
AF319045.1 |
- |
20, 21 |
3516 |
|
AK294098.1 |
- |
5, 6 |
1002 |
|
AK296532.1 |
- |
9, 10 |
1548 |
|
AK315113.1 |
- |
20, 21 |
3481 |
|
BC093780.1 |
- |
20, 21 |
3419 |
7939625 |
BC113373.1 |
- |
20, 21 |
3419 |
8322425 |
KJ534810.1 |
- |
4, 5 |
919 |
|
KJ534811.1 |
- |
19, 20 |
3265 |
|
KJ534812.1 |
- |
|
475 |
|
KJ534813.1 |
- |
7, 8 |
1375 |
|
KJ535069.1 |
- |
|
1305 |
|
KJ535088.1 |
- |
9, 10 |
1853 |
|
View Full Product Details