Gene Transcripts

Gene Symbol
CCNE1
Entrez Gene ID
898
Gene Name
cyclin E1
Gene Aliases
CCNE, pCCNE1
Chromosome Location
Chr. 19: 29811994 - 29824317 on Build GRCh38
UniGene ID
Hs.244723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001238.3 NP_001229.1 10 1052
NM_001322259.1 NP_001309188.1 9 917
NM_001322261.1 NP_001309190.1 9 905
NM_001322262.1 NP_001309191.1 9 1005
XM_011527440.2 XP_011525742.1 10 938
GenBank mRNA AK291549.1 - 10 1052
BC035498.2 - 10 1042 3841192
KF672847.1 - 7 746
KF672848.1 - 9 905
KR134287.1 - 9 917
M73812.1 - 8 881
M74093.1 - 8 849

View Full Product Details