Gene Transcripts

Gene Symbol
OR1J4
Entrez Gene ID
26219
Gene Name
olfactory receptor family 1 subfamily J member 4
Gene Aliases
HSHTPCRX01, HTPCRX01, OR9-21
Chromosome Location
Chr. 9: 122519141 - 122520082 on Build GRCh38
UniGene ID
Hs.632679
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004452.1 NP_001004452.1 1 247
GenBank mRNA BC131606.1 - 1 246

View Full Product Details