Gene Transcripts

Gene Symbol
ANKRD2
Entrez Gene ID
26287
Gene Name
ankyrin repeat domain 2
Gene Aliases
ARPP
Chromosome Location
Chr. 10: 97572441 - 97583884 on Build GRCh38
UniGene ID
Hs.73708
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129981.2 NP_001123453.1 6 991
NM_001291218.1 NP_001278147.1 6 991
NM_001291219.1 NP_001278148.1 6 991
NM_020349.3 NP_065082.2 6 991
GenBank mRNA AB058599.1 - 6 653
AJ304805.1 - 6 653
AJ583444.1 - 6 991
AK056990.1 - 6 933
AK314763.1 - 6 653
BC020817.1 - 6 653
BC107759.1 - 6 894 4096546

View Full Product Details