Gene Transcripts

Gene Symbol
GALNT8
Entrez Gene ID
26290
Gene Name
polypeptide N-acetylgalactosaminyltransferase 8
Gene Aliases
GALNAC-T8
Chromosome Location
Chr. 12: 4720586 - 4772726 on Build GRCh38
UniGene ID
Hs.511985
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017417.1 NP_059113.1 3, 4 768
GenBank mRNA AJ271385.1 - 3, 4 768
BC140888.1 - 3, 4 734 9021756
BC140889.1 - 3, 4 734 9021757

View Full Product Details