Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012430.4 | NP_036562.2 | 5 | 670 | |
XM_011512673.2 | XP_011510975.1 | 5 | 2186 | ||
XM_011512675.2 | XP_011510977.1 | 6 | 837 | ||
XM_011512676.2 | XP_011510978.1 | 5 | 726 | ||
GenBank mRNA | AF100749.1 | - | 5 | 652 | |
AK057587.1 | - | 4 | 1486 | ||
AK315771.1 | - | 5 | 639 | ||
BC007122.1 | - | 4 | 618 | 3459649 |