Gene Transcripts
- Gene Symbol
- NAAA
- Entrez Gene ID
- 27163
- Gene Name
- N-acylethanolamine acid amidase
- Gene Aliases
- ASAHL, PLT
- Chromosome Location
- Chr. 4: 75910655 - 75941013 on Build GRCh38
- UniGene ID
- Hs.437365
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042402.1 |
NP_001035861.1
|
6, 7 |
914 |
|
NM_014435.3 |
NP_055250.2
|
6, 7 |
914 |
|
XM_005262920.3 |
XP_005262977.1
|
6, 7 |
912 |
|
XM_005262921.3 |
XP_005262978.1
|
6, 7 |
912 |
|
XM_005262923.2 |
XP_005262980.1
|
6, 7 |
912 |
|
XM_005262924.2 |
XP_005262981.1
|
6, 7 |
912 |
|
XM_005262925.3 |
XP_005262982.1
|
6, 7 |
912 |
|
XM_006714180.2 |
XP_006714243.1
|
6, 7 |
912 |
|
XM_011531852.2 |
XP_011530154.1
|
6, 7 |
912 |
|
XM_011531854.2 |
XP_011530156.1
|
6, 7 |
912 |
|
XM_017008027.1 |
XP_016863516.1
|
6, 7 |
912 |
|
XM_017008028.1 |
XP_016863517.1
|
6, 7 |
912 |
|
XM_017008029.1 |
XP_016863518.1
|
6, 7 |
912 |
|
XM_017008030.1 |
XP_016863519.1
|
6, 7 |
912 |
|
GenBank mRNA |
AB161353.1 |
- |
6, 7 |
850 |
|
AK024677.1 |
- |
|
1414 |
|
AK301132.1 |
- |
5, 6 |
739 |
|
BC006388.2 |
- |
5, 6 |
817 |
4122755 |
M92449.1 |
- |
5, 6 |
610 |
|
W47597.1 |
- |
1, 2 |
171 |
|
View Full Product Details