Gene Transcripts

Gene Symbol
LSM1
Entrez Gene ID
27257
Gene Name
LSM1 homolog, mRNA degradation associated
Gene Aliases
CASM, YJL124C
Chromosome Location
Chr. 8: 38163321 - 38176730 on Build GRCh38
UniGene ID
Hs.425311
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014462.2 NP_055277.1 2, 3 519
NR_045493.1 2, 3 241
GenBank mRNA AF000177.1 - 2, 3 273
AF147348.1 - 2, 3 162
AJ238094.1 - 2, 3 297
AK312159.1 - 2, 3 272
BC001767.1 - 2, 3 206 3353833
BP376421.1 - 2, 3 482
BU941962.1 - 2, 3 244
KU178606.1 - 2, 3 109

View Full Product Details