Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290046.1 | NP_001276975.1 | 2 | 152 | |
NM_001290047.1 | NP_001276976.1 | 2 | 621 | ||
XM_011546128.2 | XP_011544430.1 | 2 | 309 | ||
XM_011546129.2 | XP_011544431.1 | 2 | 309 | ||
XM_011546132.2 | XP_011544434.1 | 2 | 309 | ||
XM_017028785.1 | XP_016884274.1 | 2 | 309 | ||
GenBank mRNA | AF336133.1 | - | 2 | 621 | |
AK308330.1 | - | 2 | 154 | ||
AL832377.1 | - | 2 | 99 | ||
BX647449.1 | - | 2 | 99 |