Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081003.2 | NP_001074472.1 | 2 | 1139 | |
NM_001081004.2 | NP_001074473.1 | 2 | 1007 | ||
NM_001287237.1 | NP_001274166.1 | 2 | 1274 | ||
NM_014066.3 | NP_054785.2 | 2 | 1161 | ||
GenBank mRNA | AF113540.1 | - | 2 | 1289 | |
BC065729.1 | - | 2 | 1129 | 6452461 |