Gene Transcripts
- Gene Symbol
- CFAP20
- Entrez Gene ID
- 29105
- Gene Name
- cilia and flagella associated protein 20
- Gene Aliases
- C16orf80, EVORF, GTL3, fSAP23
- Chromosome Location
- Chr. 16: 58113588 - 58129425 on Build GRCh38
- UniGene ID
- Hs.532755
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013242.2 |
NP_037374.1
|
1, 2 |
359 |
|
NR_136632.1 |
|
1, 2 |
392 |
|
GenBank mRNA |
AF087845.1 |
- |
1, 2 |
339 |
|
AF093680.1 |
- |
1, 2 |
338 |
|
AK027099.1 |
- |
1, 2 |
280 |
|
AK297819.1 |
- |
1, 2 |
314 |
|
AV751497.1 |
- |
1, 2 |
359 |
|
BC005152.2 |
- |
1, 2 |
328 |
3528301 |
CN364433.1 |
- |
1, 2 |
319 |
|
HY042096.1 |
- |
1, 2 |
393 |
|
View Full Product Details