Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013366.3 | NP_037498.1 | 11 | 2069 | |
GenBank mRNA | AB037827.1 | - | 9 | 1245 | |
AF191337.1 | - | 11 | 1996 | ||
BC001579.1 | - | 9 | 1261 | ||
BC009487.2 | - | 7 | 860 | ||
BC032503.1 | - | 11 | 2001 | 5243107 |