Gene Transcripts

Gene Symbol
ITSN2
Entrez Gene ID
50618
Gene Name
intersectin 2
Gene Aliases
PRO2015, SH3D1B, SH3P18, SWA, SWAP
Chromosome Location
Chr. 2: 24202866 - 24360528 on Build GRCh38
UniGene ID
Hs.432562
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006277.2 NP_006268.2 23 3133
NM_019595.3 NP_062541.3 22 3052
NM_147152.2 NP_671494.2 23 3133
GenBank mRNA AB033082.1 - 22 2959
AF001630.1 - 3 412
AF038189.1 - 3 535
AF182198.1 - 23 3112
AF182199.1 - 23 3112
AF248540.1 - 21 2833
BC146779.1 - 22 2959 9007231
U61167.1 - 1 43

View Full Product Details