Gene Transcripts

Gene Symbol
RDH8
Entrez Gene ID
50700
Gene Name
retinol dehydrogenase 8 (all-trans)
Gene Aliases
PRRDH, SDR28C2
Chromosome Location
Chr. 19: 10013249 - 10022278 on Build GRCh38
UniGene ID
Hs.675522
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015725.2 NP_056540.2 6 1483
GenBank mRNA AF229845.1 - 6 1483
AK024022.1 - 6 1248

View Full Product Details