Gene Transcripts
- Gene Symbol
- ASCC1
- Entrez Gene ID
- 51008
- Gene Name
- activating signal cointegrator 1 complex subunit 1
- Gene Aliases
- ASC1p50, CGI-18, SMABF2, p50
- Chromosome Location
- Chr. 10: 72096032 - 72217134 on Build GRCh38
- UniGene ID
- Hs.500007
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001198798.2 |
NP_001185727.1
|
6, 7 |
928 |
|
NM_001198799.2 |
NP_001185728.1
|
7, 8 |
973 |
|
NM_001198800.2 |
NP_001185729.1
|
6, 7 |
889 |
|
NR_045564.1 |
|
6, 7 |
968 |
|
XM_006717873.3 |
XP_006717936.1
|
6, 7 |
1130 |
|
XM_011539841.2 |
XP_011538143.1
|
6, 7 |
753 |
|
XM_011539842.2 |
XP_011538144.1
|
6, 7 |
1130 |
|
XM_017016291.1 |
XP_016871780.1
|
6, 7 |
753 |
|
XM_017016292.1 |
XP_016871781.1
|
6, 7 |
1130 |
|
XM_017016293.1 |
XP_016871782.1
|
6, 7 |
753 |
|
XM_017016294.1 |
XP_016871783.1
|
6, 7 |
1130 |
|
XM_017016301.1 |
XP_016871790.1
|
6, 7 |
753 |
|
XM_017016302.1 |
XP_016871791.1
|
6, 7 |
1130 |
|
GenBank mRNA |
AF132952.1 |
- |
7, 8 |
1041 |
|
AK023436.1 |
- |
6, 7 |
736 |
|
AK094170.1 |
- |
7, 8 |
807 |
|
AK096409.1 |
- |
4, 5 |
1439 |
|
AK298302.1 |
- |
6, 7 |
968 |
|
AL359339.1 |
- |
1, 2 |
47 |
|
AY013290.1 |
- |
7, 8 |
1041 |
|
BC012291.1 |
- |
6, 7 |
775 |
3858865 |
View Full Product Details