Gene Transcripts
- Gene Symbol
- CDC25A
- Entrez Gene ID
- 993
- Gene Name
- cell division cycle 25A
- Gene Aliases
- CDC25A2
- Chromosome Location
- Chr. 3: 48157178 - 48188658 on Build GRCh38
- UniGene ID
- Hs.437705
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001789.2 |
NP_001780.2
|
10, 11 |
1401 |
|
NM_201567.1 |
NP_963861.1
|
9, 10 |
1281 |
|
XM_006713434.2 |
XP_006713497.1
|
10, 11 |
1493 |
|
XM_006713435.2 |
XP_006713498.1
|
9, 10 |
1373 |
|
XM_011534316.1 |
XP_011532618.1
|
11, 12 |
1594 |
|
GenBank mRNA |
AK290554.1 |
- |
10, 11 |
1076 |
|
AY137580.1 |
- |
9, 10 |
917 |
|
BC007401.2 |
- |
10, 11 |
1401 |
2989279 |
BC018642.2 |
- |
10, 11 |
1401 |
3687561 |
EU812118.1 |
- |
9, 10 |
914 |
|
FJ866789.1 |
- |
10, 11 |
1034 |
|
M81933.1 |
- |
10, 11 |
1493 |
|
View Full Product Details