Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304722.1 | NP_001291651.1 | 5 | 759 | |
NM_001304723.1 | NP_001291652.1 | 4 | 732 | ||
NM_016274.5 | NP_057358.2 | 5 | 792 | ||
XM_017001420.1 | XP_016856909.1 | 5 | 769 | ||
GenBank mRNA | AF073836.1 | - | 4 | 422 | |
AF168676.1 | - | 5 | 524 | ||
AF192912.1 | - | 5 | 729 | ||
AF217956.1 | - | 5 | 580 | ||
AF291105.1 | - | 5 | 720 | ||
AK025937.1 | - | 3 | 209 | ||
AK125609.1 | - | 3 | 2824 | ||
AK222921.1 | - | 5 | 436 | ||
AK302255.1 | - | 5 | 776 | ||
BC010149.2 | - | 5 | 520 | 4330014 | |
BP329810.1 | - | 4 | 481 | ||
BX353413.2 | - | 5 | 760 | ||
BX376480.2 | - | 5 | 728 | ||
BX394578.2 | - | 5 | 658 |