Gene Transcripts

Gene Symbol
GP6
Entrez Gene ID
51206
Gene Name
glycoprotein VI platelet
Gene Aliases
BDPLT11, GPIV, GPVI
Chromosome Location
Chr. 19: 55013705 - 55038264 on Build GRCh38
UniGene ID
Hs.661752
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083899.2 NP_001077368.2 5, 6 692
NM_016363.5 NP_057447.5 5, 6 692

View Full Product Details