Gene Transcripts

Gene Symbol
EMC4
Entrez Gene ID
51234
Gene Name
ER membrane protein complex subunit 4
Gene Aliases
PIG17, TMEM85
Chromosome Location
Chr. 15: 34224997 - 34230165 on Build GRCh38
UniGene ID
Hs.250905
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016454.3 NP_057538.1 4 522
GenBank mRNA AF151018.1 - 4 475
AK075227.1 - 4 501
AY336092.1 - 4 462
KU178641.1 - 4 404

View Full Product Details