Gene Transcripts

Gene Symbol
THEG
Entrez Gene ID
51298
Gene Name
theg spermatid protein
Gene Aliases
CT56, THEG1
Chromosome Location
Chr. 19: 362057 - 376693 on Build GRCh38
UniGene ID
Hs.250002
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016585.4 NP_057669.1 6 764
NM_199202.2 NP_954672.1 5 692
XM_011528049.2 XP_011526351.1 6 1443
XM_011528050.2 XP_011526352.1 5 1370
XM_011528051.2 XP_011526353.1 3 1240
GenBank mRNA AB033129.1 - 6 764
AF268610.1 - 6 730
BC028574.2 - 6 743 4821678

View Full Product Details