Gene Transcripts
- Gene Symbol
- REEP2
- Entrez Gene ID
- 51308
- Gene Name
- receptor accessory protein 2
- Gene Aliases
- C5orf19, SGC32445, SPG72, Yip2d
- Chromosome Location
- Chr. 5: 138439001 - 138446969 on Build GRCh38
- UniGene ID
- Hs.416090
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271803.1 |
NP_001258732.1
|
6, 7 |
767 |
|
NM_016606.3 |
NP_057690.2
|
6, 7 |
761 |
|
NR_073448.1 |
|
6, 7 |
836 |
|
NR_073449.1 |
|
6, 7 |
842 |
|
GenBank mRNA |
AF070551.1 |
- |
2, 3 |
205 |
|
AF251041.1 |
- |
5, 6 |
541 |
|
AK056193.1 |
- |
6, 7 |
723 |
|
AK223611.1 |
- |
6, 7 |
708 |
|
AK290547.1 |
- |
6, 7 |
657 |
|
AK291101.1 |
- |
6, 7 |
720 |
|
AK293480.1 |
- |
6, 7 |
735 |
|
AY562240.1 |
- |
6, 7 |
553 |
|
BC006218.2 |
- |
6, 7 |
675 |
3940709 |
View Full Product Details