Gene Transcripts

Gene Symbol
TRAPPC2L
Entrez Gene ID
51693
Gene Name
trafficking protein particle complex 2-like
Gene Aliases
HSPC176
Chromosome Location
Chr. 16: 88856220 - 88862686 on Build GRCh38
UniGene ID
Hs.461722
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318524.1 NP_001305453.1 3 303
NM_001318525.1 NP_001305454.1 3 303
NM_001318526.1 NP_001305455.1 3 368
NM_001318527.1 NP_001305456.1 3 619
NM_001318528.1 NP_001305457.1 3 280
NM_001318529.1 NP_001305458.1 3 619
NM_001318530.1 NP_001305459.1 3 280
NM_001318532.1 NP_001305461.1 3 619
NM_016209.4 NP_057293.1 3 303
NR_134670.1 4 490
NR_134671.1 4 419
XM_017023270.1 XP_016878759.1 3 350
GenBank mRNA AF089106.1 - 3 296
AF161524.1 - 3 258
AK126779.1 - 3 262
AK307253.1 - 4 419
AK310805.1 - 3 574
AK311885.1 - 3 272
AX771012.1 - 3 296
AX771013.1 - 3 258
BC011369.1 - 3 250 3868798
BC018024.1 - 3 257 4287920
BC105809.1 - 3 269 6745065
BI754306.1 - 4 432
BX340702.2 - 3 565
BX384775.2 - 3 574
BX419849.2 - 3 248
DA209193.1 - 3 280
DA293122.1 - 3 303
DA295904.1 - 3 239
DA833474.1 - 3 242

View Full Product Details