Gene Transcripts
- Gene Symbol
- GDAP1
- Entrez Gene ID
- 54332
- Gene Name
- ganglioside induced differentiation associated protein 1
- Gene Aliases
- CMT4, CMT4A, CMTRIA
- Chromosome Location
- Chr. 8: 74349251 - 74488867 on Build GRCh38
- UniGene ID
- Hs.168950
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001040875.2 |
NP_001035808.1
|
4, 5 |
522 |
|
NM_018972.2 |
NP_061845.2
|
4, 5 |
652 |
|
NR_046346.1 |
|
4, 5 |
507 |
|
XM_011517551.2 |
XP_011515853.1
|
3, 4 |
1610 |
|
XM_011517552.2 |
XP_011515854.1
|
3, 4 |
447 |
|
XM_017013585.1 |
XP_016869074.1
|
4, 5 |
1784 |
|
GenBank mRNA |
AB551556.1 |
- |
4, 5 |
651 |
|
AB551557.1 |
- |
4, 5 |
651 |
|
AB551558.1 |
- |
4, 5 |
649 |
|
AK292572.1 |
- |
4, 5 |
591 |
|
AK295594.1 |
- |
4, 5 |
446 |
|
BC024939.1 |
- |
4, 5 |
487 |
3923335 |
BC035450.1 |
- |
4, 5 |
499 |
|
Y17849.1 |
- |
4, 5 |
580 |
|
View Full Product Details