Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018990.3 | NP_061863.1 | 2 | 318 | |
XM_006724763.1 | XP_006724826.1 | 2 | 285 | ||
GenBank mRNA | AK292023.1 | - | 2 | 266 | |
AK314869.1 | - | 2 | 285 | ||
AL049683.1 | - | 2 | 243 | ||
BC051881.1 | - | 2 | 250 | 6011968 | |
CR975883.1 | - | 2 | 318 |